Assay Details
| Gene Symbol: Sh3gl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019535.2 | NP_062408.2 | 5 - 6 | 584 | 102 | |
| GenBank mRNA | AF326561.1 | - | 5 - 6 | 463 | 102 | |
| AF326562.1 | - | 5 - 6 | 463 | 102 | ||
| AK035210.1 | - | 915 | 102 | |||
| AK043424.1 | - | 572 | 102 | |||
| AK044370.1 | - | 5 - 6 | 599 | 102 | ||
| AK134970.1 | - | 3 - 4 | 295 | 102 | ||
| AK139337.1 | - | 5 - 6 | 549 | 102 | ||
| BC018385.1 | - | 5 - 6 | 584 | 4503038 | 102 | |
| U58886.1 | - | 5 - 6 | 547 | 102 | ||
More Information
Set Membership: |
|

Sh3gl2
Genomic Map