Assay Details
| Gene Symbol: Cdh13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019707.4 | NP_062681.2 | 10 - 11 | 1635 | 61 | |
| GenBank mRNA | AB022100.1 | - | 10 - 11 | 1644 | 61 | |
| AK028002.1 | - | 2 - 3 | 447 | 61 | ||
| AK032409.1 | - | 10 - 11 | 1666 | 61 | ||
| AK039438.1 | - | 10 - 11 | 1657 | 61 | ||
| AK048724.1 | - | 10 - 11 | 1635 | 61 | ||
| AK134649.1 | - | 10 - 11 | 1594 | 61 | ||
More Information
Set Membership: |
|

Cdh13
Genomic Map