Assay Details
| Gene Symbol: Snd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019776.2 | NP_062750.2 | 17 - 18 | 2182 | 59 | |
| GenBank mRNA | AB021491.1 | - | 17 - 18 | 2139 | 59 | |
| AK088028.1 | - | 17 - 18 | 2141 | 59 | ||
| AK133837.1 | - | 17 - 18 | 2188 | 59 | ||
| AK159837.1 | - | 17 - 18 | 2147 | 59 | ||
| AK161591.1 | - | 17 - 18 | 2136 | 59 | ||
| AK162432.1 | - | 17 - 18 | 2123 | 59 | ||
| BC007126.1 | - | 17 - 18 | 2129 | 3592548 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Snd1
Genomic Map