Assay Details
| Gene Symbol: Maged1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019791.2 | NP_062765.1 | 2 - 3 | 255 | 92 | |
| GenBank mRNA | AB029448.1 | - | 2 - 3 | 255 | 92 | |
| AK017275.1 | - | 2 - 3 | 173 | 92 | ||
| AK049660.1 | - | 2 - 3 | 206 | 92 | ||
| AK087937.1 | - | 2 - 3 | 206 | 92 | ||
| AK132248.1 | - | 2 - 3 | 194 | 92 | ||
| AK135452.1 | - | 2 - 3 | 205 | 92 | ||
| AK145872.1 | - | 2 - 3 | 175 | 92 | ||
| AK146867.1 | - | 2 - 3 | 205 | 92 | ||
| AK146908.1 | - | 2 - 3 | 205 | 92 | ||
| AK220150.1 | - | 2 - 3 | 184 | 92 | ||
| BC016438.1 | - | 2 - 3 | 171 | 4480947 | 92 | |
| BC031461.1 | - | 2 - 3 | 195 | 4018875 | 92 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB029448.1,AK017275.1,AK049660.1,AK132248.1,AK135452.1,AK145872.1,AK146867.1,AK146908.1,BC016438.1,BC031461.1,NM_019791.2 transcripts.
Set Membership: |
|

Maged1
Genomic Map