Assay Details
| Gene Symbol: Maged1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019791.2 | NP_062765.1 | 11 - 12 | 2161 | 75 | |
| GenBank mRNA | AB029448.1 | - | 11 - 12 | 2161 | 75 | |
| AF319975.1 | - | 10 - 11 | 1931 | 75 | ||
| AK013231.1 | - | 3 - 4 | 684 | 74 | ||
| AK017275.1 | - | 11 - 12 | 2079 | 75 | ||
| AK049660.1 | - | 11 - 12 | 2112 | 75 | ||
| AK087937.1 | - | 11 - 12 | 2111 | 75 | ||
| AK132248.1 | - | 11 - 12 | 2100 | 75 | ||
| AK135452.1 | - | 11 - 12 | 2111 | 75 | ||
| AK145872.1 | - | 11 - 12 | 2081 | 75 | ||
| AK146867.1 | - | 11 - 12 | 2111 | 75 | ||
| AK146908.1 | - | 11 - 12 | 2111 | 75 | ||
| AK150876.1 | - | 8 - 9 | 1100 | 75 | ||
| AK220150.1 | - | 12 - 13 | 2018 | 75 | ||
| BC016438.1 | - | 10 - 11 | 2295 | 4480947 | 75 | |
| BC031461.1 | - | 11 - 12 | 2101 | 4018875 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Maged1
Genomic Map