Assay Details
| Gene Symbol: Ccnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019937.3 | NP_064321.2 | 4 - 5 | 720 | 89 | |
| GenBank mRNA | AF159159.1 | - | 4 - 5 | 673 | 89 | |
| AF467251.1 | - | 4 - 5 | 714 | 89 | ||
| AK011629.1 | - | 4 - 5 | 677 | 89 | ||
| BC007177.1 | - | 5 - 6 | 767 | 89 | ||
| BC037204.1 | - | 4 - 5 | 658 | 89 | ||
| BC069909.1 | - | 5 - 6 | 804 | 89 | ||
| BC094383.1 | - | 4 - 5 | 656 | 30604933 | 89 | |
| BC117816.1 | - | 5 - 6 | 613 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccnl1
Genomic Map