Assay Details
| Gene Symbol: Ccnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019937.3 | NP_064321.2 | 7 - 8 | 999 | 103 | |
| GenBank mRNA | AF159159.1 | - | 7 - 8 | 952 | 103 | |
| AF467251.1 | - | 7 - 8 | 993 | 103 | ||
| BC007177.1 | - | 7 - 8 | 2735 | 103 | ||
| BC037204.1 | - | 6 - 7 | 2626 | 103 | ||
| BC069909.1 | - | 8 - 9 | 1083 | 103 | ||
| BC094383.1 | - | 7 - 8 | 935 | 30604933 | 103 | |
| BC117816.1 | - | 8 - 9 | 892 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccnl1
Genomic Map