Assay Details
| Gene Symbol: Mpp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164733.1 | NP_001158205.1 | 2 - 3 | 282 | 98 | |
| NM_001164734.1 | NP_001158206.1 | 2 - 3 | 292 | 98 | ||
| NM_019939.2 | NP_064323.2 | 2 - 3 | 292 | 98 | ||
| GenBank mRNA | AF161181.1 | - | 2 - 3 | 156 | 98 | |
| AF199009.1 | - | 2 - 3 | 288 | 98 | ||
| AF199010.1 | - | 2 - 3 | 288 | 98 | ||
| AK144434.1 | - | 2 - 3 | 293 | 98 | ||
| AK164549.1 | - | 2 - 3 | 283 | 98 | ||
| BC042868.2 | - | 2 - 3 | 239 | 98 | ||
| BC046421.1 | - | 2 - 3 | 239 | 98 | ||
| BC069908.1 | - | 2 - 3 | 281 | 98 | ||
| BC106140.1 | - | 2 - 3 | 239 | 98 | ||
| BC138666.1 | - | 2 - 3 | 150 | 8861687 | 98 | |
| BC145363.1 | - | 2 - 3 | 150 | 9053921 | 98 | |
| BC145364.1 | - | 2 - 3 | 140 | 9053922 | 98 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF161181.1,AF199009.1,AF199010.1,AK144434.1,AK164549.1,BC046421.1,BC069908.1,BC106140.1,BC138666.1,BC145363.1,BC145364.1,NM_001164733.1,NM_001164734.1,NM_019939.2 transcripts.
Set Membership: |
|

Mpp6
Genomic Map