Assay Details
| Gene Symbol: Mpp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164733.1 | NP_001158205.1 | 10 - 11 | 1270 | 131 | |
| NM_001164734.1 | NP_001158206.1 | 10 - 11 | 1280 | 131 | ||
| NM_019939.2 | NP_064323.2 | 9 - 10 | 1238 | 131 | ||
| GenBank mRNA | AF161181.1 | - | 9 - 10 | 1102 | 131 | |
| AF199009.1 | - | 9 - 10 | 1234 | 131 | ||
| AF199010.1 | - | 10 - 11 | 1276 | 131 | ||
| AK144434.1 | - | 10 - 11 | 1281 | 131 | ||
| AK178596.1 | - | 4 - 5 | 357 | 131 | ||
| AK187690.1 | - | 1 - 2 | 78 | 131 | ||
| BC138666.1 | - | 9 - 10 | 1096 | 8861687 | 131 | |
| BC145363.1 | - | 10 - 11 | 1138 | 9053921 | 131 | |
| BC145364.1 | - | 10 - 11 | 1128 | 9053922 | 131 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mpp6
Genomic Map