Assay Details
| Gene Symbol: Stat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019963.1 | NP_064347.1 | 15 - 16 | 1492 | 85 | |
| GenBank mRNA | AF187231.1 | - | 15 - 16 | 1489 | 85 | |
| AF206162.1 | - | 15 - 16 | 1492 | 85 | ||
| AK030903.1 | - | 8 - 9 | 584 | 85 | ||
| AK149924.1 | - | 15 - 16 | 1505 | 85 | ||
| AK150070.1 | - | 15 - 16 | 1504 | 85 | ||
| AK153405.1 | - | 13 - 14 | 1139 | 85 | ||
| BC064827.1 | - | 4 - 5 | 489 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC064827.1 transcripts.
Set Membership: |
|

Stat2
Genomic Map