Assay Details
| Gene Symbol: Sort1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271599.1 | NP_001258528.1 | 5 - 6 | 785 | 112 | |
| NM_019972.3 | NP_064356.2 | 5 - 6 | 785 | 112 | ||
| GenBank mRNA | AF175279.1 | - | 5 - 6 | 714 | 112 | |
| AK147442.1 | - | 5 - 6 | 768 | 112 | ||
| BC034129.1 | - | 2 - 3 | 218 | 112 | ||
| BC056343.1 | - | 5 - 6 | 717 | 5704346 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sort1
Genomic Map