Assay Details
| Gene Symbol: Arhgap31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020260.2 | NP_064656.2 | 4 - 5 | 787 | 78 | |
| GenBank mRNA | AF151363.1 | - | 4 - 5 | 428 | 78 | |
| AK018057.1 | - | 4 - 5 | 653 | 78 | ||
| AK138417.1 | - | 4 - 5 | 1027 | 78 | ||
| AK149975.1 | - | 4 - 5 | 623 | 78 | ||
| AK150109.1 | - | 4 - 5 | 623 | 78 | ||
| AK155729.1 | - | 4 - 5 | 607 | 78 | ||
| AK170685.1 | - | 4 - 5 | 607 | 78 | ||
| BC138870.1 | - | 4 - 5 | 592 | 8861892 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap31
Genomic Map