Assay Details
| Gene Symbol: Arhgap31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020260.2 | NP_064656.2 | 9 - 10 | 1414 | 93 | |
| GenBank mRNA | AF151363.1 | - | 9 - 10 | 1055 | 93 | |
| AK138417.1 | - | 9 - 10 | 1654 | 93 | ||
| AK149975.1 | - | 9 - 10 | 1250 | 93 | ||
| AK150109.1 | - | 9 - 10 | 1250 | 93 | ||
| AK155729.1 | - | 9 - 10 | 1234 | 93 | ||
| AK170685.1 | - | 9 - 10 | 1234 | 93 | ||
| BC138870.1 | - | 9 - 10 | 1219 | 8861892 | 93 | |
More Information
Set Membership: |
|

Arhgap31
Genomic Map