Assay Details
Gene Symbol: Ctsr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020284.1 | NP_064680.1 | 7 - 8 | 976 | 72 | |
GenBank mRNA | AF245399.1 | - | 7 - 8 | 976 | 72 | |
AK005429.1 | - | 7 - 8 | 996 | 72 | ||
AK014432.1 | - | 7 - 8 | 992 | 72 | ||
AY014778.1 | - | 7 - 8 | 935 | 72 | ||
AY943668.1 | - | 7 - 8 | 1048 | 72 | ||
BC100338.1 | - | 7 - 8 | 971 | 30712889 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P272806 | BC100338.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P272806 | AY014778.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P272806 | AF245399.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P272806 | AK005429.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P272806 | AK014432.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P272806 | BC100338.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P272806 | AY014778.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P272806 | AF245399.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P272806 | AK005429.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P272806 | AK014432.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 219000 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 707668 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 288477 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133148 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P272806 | BC100338.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P272806 | AY014778.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P272806 | AF245399.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P272806 | AK005429.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P272806 | AK014432.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P272806 | BC100338.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P272806 | AY014778.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P272806 | AF245399.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P272806 | AK005429.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P272806 | AK014432.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P272806 | BC100338.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P272806 | AY014778.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P272806 | AF245399.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P272806 | AK005429.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P272806 | AK014432.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P272806 | BC100338.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P272806 | AY014778.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P272806 | AF245399.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P272806 | AK005429.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P272806 | AK014432.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088823 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088818 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088812 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088831 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088817 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088821 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088841 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088815 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088824 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088825 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088830 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088828 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088839 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088840 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088822 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088842 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088832 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088836 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088816 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088834 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088829 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088826 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088814 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088835 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07896848 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133148 | - | - |
Whole Mouse Genome G4122A | A_51_P272806 | BC100338.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P272806 | AY014778.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P272806 | AF245399.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P272806 | AK005429.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P272806 | AK014432.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P272806 | BC100338.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P272806 | AY014778.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P272806 | AF245399.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P272806 | AK005429.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P272806 | AK014432.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103512 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133148 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10863026 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10863029 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10511615 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10863019 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10863030 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10863021 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10863027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088803 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088813 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088835 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088809 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088834 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088851 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088794 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088795 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088810 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088830 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088787 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088831 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088818 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088828 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088798 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088807 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088824 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088817 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088839 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088797 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088845 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088841 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088844 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088815 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088829 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088859 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088812 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088785 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088823 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088821 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088814 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088826 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088816 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088855 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088801 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088847 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088805 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088792 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088832 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088850 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088842 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088814 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088851 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088797 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088834 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088803 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088847 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088837 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088794 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088822 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10441340 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088807 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088818 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088844 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088798 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088801 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088845 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10644758 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088823 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088840 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10974709 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088790 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088821 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088813 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088830 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088791 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088848 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088853 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10511615 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088785 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088809 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10863019 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088828 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088852 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10677492 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10441335 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088815 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088841 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088805 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088795 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088835 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10863024 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088831 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10684098 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088826 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10863035 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088812 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088836 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088792 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088850 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088787 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10409912 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10409909 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10409910 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10409908 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10409907 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10409906 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10409911 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10409913 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1419142_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10409905 | - | - |
Mouse Genome 430 2.0 Array | 1419142_at | - | - |
Mouse Genome 430 2.0 Array | 1419142_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56835_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56835_at | - | - |
Mouse Expression Set 430 | 1419142_PM_at | - | - |
Mouse Expression Set 430 | 1419142_at | - | - |
Mouse Genome 430A 2.0 Array | 1419142_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1419142_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419142_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419142_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 3830433 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003830433 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2712480 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2712480 | - | - |
mouse-6 v1.1 (Illumina) | scl43859.8.1_132-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3830433 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl43859.8.1_132-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3830433 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2712480 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2712480 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl43859.8.1_132-S | - | - |
