Assay Details
| Gene Symbol: Srf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020493.2 | NP_065239.1 | 5 - 6 | 1678 | 80 | |
| GenBank mRNA | BC051950.1 | - | 5 - 6 | 1691 | 6404515 | 80 |
More Information
Updated Mapping Information
Set Membership: |
|

Srf
Genomic Map