Assay Details
| Gene Symbol: As3mt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020577.2 | NP_065602.2 | 3 - 4 | 263 | 71 | |
| GenBank mRNA | AF166383.1 | - | 3 - 4 | 233 | 71 | |
| AK002801.1 | - | 3 - 4 | 196 | 71 | ||
| AK006571.1 | - | 3 - 4 | 248 | 71 | ||
| AK011320.1 | - | 3 - 4 | 215 | 71 | ||
| BC013468.1 | - | 3 - 4 | 188 | 71 | ||
| BC145663.1 | - | 3 - 4 | 205 | 40130966 | 71 | |
| BY287843.1 | - | 3 - 4 | 263 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

As3mt
Genomic Map