Assay Details
| Gene Symbol: As3mt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020577.2 | NP_065602.2 | 4 - 5 | 420 | 97 | |
| GenBank mRNA | AF166383.1 | - | 4 - 5 | 390 | 97 | |
| AK002801.1 | - | 4 - 5 | 353 | 97 | ||
| AK006571.1 | - | 4 - 5 | 405 | 97 | ||
| AK009814.1 | - | 3 - 4 | 407 | 97 | ||
| AK011320.1 | - | 4 - 5 | 372 | 97 | ||
| BC013468.1 | - | 4 - 5 | 97 | |||
| BC145663.1 | - | 4 - 5 | 40130966 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

As3mt
Genomic Map