Assay Details
| Gene Symbol: Ces1g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021456.4 | NP_067431.2 | 6 - 7 | 851 | 100 | |
| GenBank mRNA | AK136446.1 | - | 6 - 7 | 848 | 100 | |
| AK136588.1 | - | 6 - 7 | 848 | 100 | ||
| AK149446.1 | - | 6 - 7 | 848 | 100 | ||
| AK162328.1 | - | 6 - 7 | 826 | 100 | ||
| BC021150.1 | - | 6 - 7 | 870 | 4163576 | 100 | |
| BC026897.1 | - | 6 - 7 | 831 | 4195247 | 100 | |
| Y12887.1 | - | 6 - 7 | 837 | 100 | ||
More Information
Set Membership: |
|

Ces1g
Genomic Map