Assay Details
| Gene Symbol: F12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC037085.1 | - | 11 - 12 | 1351 | 75 | |
| BC049867.1 | - | 11 - 12 | 1374 | 75 | ||
| BC057921.1 | - | 11 - 12 | 1362 | 5101105 | 75 | |
| X99571.1 | - | 11 - 12 | 1357 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

F12
Genomic Map