Assay Details
| Gene Symbol: Slc4a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021530.2 | NP_067505.2 | 20 - 21 | 2944 | 75 | |
| GenBank mRNA | AF224508.1 | - | 20 - 21 | 2890 | 75 | |
| AK034237.1 | - | 20 - 21 | 2946 | 75 | ||
| AK039404.1 | - | 20 - 21 | 2909 | 75 | ||
| AK047863.1 | - | 13 - 14 | 1878 | 75 | ||
| AK171610.1 | - | 19 - 20 | 2720 | 75 | ||
| AK173014.1 | - | 20 - 21 | 2778 | 75 | ||
| BC030388.1 | - | 20 - 21 | 2906 | 5401143 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc4a8
Genomic Map