Assay Details
| Gene Symbol: Carm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021531.6 | NP_067506.2 | 8 - 9 | 1141 | 77 | |
| NM_153141.1 | NP_694781.1 | 8 - 9 | 1141 | 77 | ||
| GenBank mRNA | AF117887.2 | - | 8 - 9 | 1051 | 77 | |
| AK158242.1 | - | 8 - 9 | 1106 | 77 | ||
| AK158757.1 | - | 8 - 9 | 981 | 77 | ||
| BC003964.1 | - | 3 - 4 | 248 | 77 | ||
| BC036974.1 | - | 8 - 9 | 1125 | 4935077 | 77 | |
| BC071267.1 | - | 9 - 10 | 1257 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Carm1
Genomic Map