Assay Details
| Gene Symbol: Midn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB036882.1 | - | 2 - 3 | 694 | 64 | |
| AK154535.1 | - | 1 - 2 | 638 | 64 | ||
| AK164324.1 | - | 2 - 3 | 712 | 64 | ||
| AK164651.1 | - | 2 - 3 | 714 | 64 | ||
| BC034719.1 | - | 2 - 3 | 664 | 3710931 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Midn
Genomic Map