Assay Details
| Gene Symbol: Midn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB036882.1 | - | 5 - 6 | 976 | 65 | |
| AK154535.1 | - | 4 - 5 | 920 | 65 | ||
| BC034719.1 | - | 5 - 6 | 946 | 3710931 | 65 | |
More Information
Set Membership: |
|

Midn
Genomic Map