Assay Details
| Gene Symbol: Sap30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021788.2 | NP_068560.1 | 2 - 3 | 732 | 94 | |
| GenBank mRNA | AF075136.1 | - | 2 - 3 | 743 | 94 | |
| AK010928.1 | - | 2 - 3 | 733 | 94 | ||
| AK088745.1 | - | 2 - 3 | 737 | 94 | ||
| BC132081.1 | - | 2 - 3 | 715 | 40130358 | 94 | |
| BC132087.1 | - | 2 - 3 | 715 | 40130364 | 94 | |
| CO800093.1 | - | 2 - 3 | 94 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Sap30
Genomic Map