Assay Details
| Gene Symbol: Map3k11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022012.3 | NP_071295.2 | 5 - 6 | 1800 | 79 | |
| GenBank mRNA | AK159518.1 | - | 3 - 4 | 820 | 79 | |
| BC030928.1 | - | 3 - 4 | 353 | 79 | ||
| BC047152.1 | - | 3 - 4 | 1547 | 6476975 | 79 | |
| BC095963.1 | - | 5 - 6 | 1800 | 5400993 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Map3k11
Genomic Map