Assay Details
| Gene Symbol: Map3k11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022012.3 | NP_071295.2 | 6 - 7 | 1977 | 76 | |
| GenBank mRNA | AK159518.1 | - | 4 - 5 | 997 | 76 | |
| BC030928.1 | - | 4 - 5 | 530 | 76 | ||
| BC047152.1 | - | 4 - 5 | 1724 | 6476975 | 76 | |
| BC095963.1 | - | 6 - 7 | 1977 | 5400993 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Map3k11
Genomic Map