Assay Details
| Gene Symbol: Sv2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022030.3 | NP_071313.1 | 7 - 8 | 1718 | 94 | |
| GenBank mRNA | AK028318.1 | - | 7 - 8 | 1663 | 94 | |
| AK080502.1 | - | 7 - 8 | 1671 | 94 | ||
| AK080525.1 | - | 7 - 8 | 1760 | 94 | ||
| AK122360.1 | - | 7 - 8 | 1754 | 94 | ||
| AK144166.1 | - | 7 - 8 | 1741 | 94 | ||
| BC046587.1 | - | 7 - 8 | 1718 | 6493509 | 94 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK158936.1 | - | Chr. - - [-] on Build | 94 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Sv2a
Genomic Map