Assay Details
| Gene Symbol: Smoc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146217.1 | NP_001139689.1 | 2 - 3 | 513 | 58 | |
| NM_022316.2 | NP_071711.2 | 2 - 3 | 513 | 58 | ||
| GenBank mRNA | AF070470.1 | - | 2 - 3 | 489 | 58 | |
| AK040931.1 | - | 2 - 3 | 514 | 58 | ||
| BC031804.1 | - | 2 - 3 | 493 | 3673732 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smoc1
Genomic Map