Assay Details
| Gene Symbol: Smoc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022316.2 | NP_071711.2 | 5 - 6 | 778 | 71 | |
| GenBank mRNA | AF070470.1 | - | 5 - 6 | 754 | 71 | |
| BC031804.1 | - | 5 - 6 | 758 | 3673732 | 71 | |
More Information
Set Membership: |
|

Smoc1
Genomic Map