Assay Details
| Gene Symbol: St7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289624.1 | NP_001276553.1 | 7 - 8 | 934 | 99 | |
| NM_001289625.1 | NP_001276554.1 | 7 - 8 | 934 | 99 | ||
| NM_001289627.1 | NP_001276556.1 | 7 - 8 | 668 | 99 | ||
| NM_022332.3 | NP_071727.2 | 7 - 8 | 1010 | 99 | ||
| GenBank mRNA | AF234884.1 | - | 7 - 8 | 961 | 99 | |
| AK079107.1 | - | 7 - 8 | 1011 | 99 | ||
| AY007792.1 | - | 7 - 8 | 896 | 99 | ||
| AY007794.1 | - | 8 - 9 | 925 | 99 | ||
| AY007796.1 | - | 8 - 9 | 735 | 99 | ||
| AY007798.1 | - | 7 - 8 | 668 | 99 | ||
| AY551821.1 | - | 7 - 8 | 661 | 99 | ||
| BC024652.1 | - | 7 - 8 | 934 | 3660309 | 99 | |
| BC108418.1 | - | 7 - 8 | 954 | 30603484 | 99 | |
More Information
Set Membership: |
|

St7
Genomic Map