Assay Details
| Gene Symbol: Rgs18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022881.4 | NP_075019.1 | 2 - 3 | 404 | 68 | |
| GenBank mRNA | AB042807.1 | - | 2 - 3 | 419 | 68 | |
| AF302685.1 | - | 2 - 3 | 410 | 68 | ||
| AK036407.1 | - | 2 - 3 | 405 | 68 | ||
| BC099387.1 | - | 2 - 3 | 399 | 30846527 | 68 | |
| BC132544.1 | - | 2 - 3 | 405 | 40130821 | 68 | |
| BC132546.1 | - | 2 - 3 | 405 | 40130823 | 68 | |
More Information
Set Membership: |
|

Rgs18
Genomic Map