Assay Details
| Gene Symbol: Dpysl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023047.2 | NP_075534.1 | 11 - 12 | 1720 | 109 | |
| GenBank mRNA | AF249295.1 | - | 11 - 12 | 1884 | 109 | |
| AK044866.1 | - | 11 - 12 | 1594 | 109 | ||
| BC065054.1 | - | 11 - 12 | 1720 | 5709296 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dpysl5
Genomic Map