Assay Details
| Gene Symbol: Sigirr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023059.3 | NP_075546.2 | 8 - 9 | 1474 | 80 | |
| GenBank mRNA | AF113795.1 | - | 7 - 8 | 991 | 80 | |
| AF239957.1 | - | 7 - 8 | 985 | 80 | ||
| AK133518.1 | - | 8 - 9 | 1099 | 80 | ||
| AK145889.1 | - | 8 - 9 | 1477 | 80 | ||
| BC010806.1 | - | 7 - 8 | 996 | 4240425 | 80 | |
| BC094069.1 | - | 8 - 9 | 1080 | 4974962 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sigirr
Genomic Map