Assay Details
| Gene Symbol: Renbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164704.1 | NP_001158176.1 | 6 - 7 | 752 | 94 | |
| NM_023132.3 | NP_075621.3 | 6 - 7 | 794 | 94 | ||
| GenBank mRNA | AK014347.1 | - | 6 - 7 | 714 | 94 | |
| AK146652.1 | - | 6 - 7 | 759 | 94 | ||
| AK167648.1 | - | 6 - 7 | 665 | 94 | ||
| AK182381.1 | - | 1 - 2 | 156 | 94 | ||
| AK194143.1 | - | 1 - 2 | 147 | 94 | ||
| AK205945.1 | - | 1 - 2 | 157 | 95 | ||
| AK215192.1 | - | 1 - 2 | 145 | 94 | ||
| AY183138.1 | - | 6 - 7 | 762 | 94 | ||
| BC004830.1 | - | 3 - 4 | 1282 | 94 | ||
| BC014821.1 | - | 6 - 7 | 688 | 4238244 | 94 | |
| BC034846.1 | - | 2 - 3 | 1478 | 94 | ||
| BC035455.1 | - | 6 - 7 | 688 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Renbp
Genomic Map