Assay Details
| Gene Symbol: Cblc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161844.1 | NP_001155316.1 | 4 - 5 | 806 | 59 | |
| NM_023224.5 | NP_075713.3 | 4 - 5 | 806 | 59 | ||
| GenBank mRNA | AF319956.1 | - | 4 - 5 | 779 | 59 | |
| AK010228.1 | - | 4 - 5 | 807 | 59 | ||
| BC046337.1 | - | 4 - 5 | 782 | 6395806 | 59 | |
| BC111037.1 | - | 4 - 5 | 790 | 5353413 | 59 | |
| BE198430.1 | - | 2 - 3 | 149 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cblc
Genomic Map