Assay Details
| Gene Symbol: Snx9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025664.5 | NP_079940.2 | 7 - 8 | 862 | 89 | |
| GenBank mRNA | AK012520.1 | - | 2 - 3 | 154 | 89 | |
| AK139802.1 | - | 7 - 8 | 896 | 89 | ||
| AK149739.1 | - | 7 - 8 | 876 | 89 | ||
| AK149849.1 | - | 7 - 8 | 916 | 89 | ||
| AK155830.1 | - | 897 | 89 | |||
| AK157517.1 | - | 7 - 8 | 878 | 89 | ||
| AK170339.1 | - | 878 | 89 | |||
| BC014814.1 | - | 7 - 8 | 862 | 3257422 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Snx9
Genomic Map