Assay Details
| Gene Symbol: Smarcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025891.3 | NP_080167.3 | 6 - 7 | 823 | 73 | |
| GenBank mRNA | AK005094.1 | - | 6 - 7 | 770 | 73 | |
| AK158019.1 | - | 793 | 73 | |||
| BC013122.1 | - | 6 - 7 | 860 | 3586276 | 73 | |
| BC060525.1 | - | 6 - 7 | 722 | 6817219 | 73 | |
More Information
Set Membership: |
|

Smarcd3
Genomic Map