Assay Details
| Gene Symbol: Mrpl45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025927.4 | NP_080203.1 | 6 - 7 | 815 | 65 | |
| GenBank mRNA | AK011163.1 | - | 6 - 7 | 684 | 65 | |
| AK011605.1 | - | 6 - 7 | 674 | 65 | ||
| AK081922.1 | - | 6 - 7 | 676 | 65 | ||
| AK157771.1 | - | 6 - 7 | 677 | 65 | ||
| AK187738.1 | - | 1 - 2 | 150 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mrpl45
Genomic Map