Assay Details
| Gene Symbol: Tpsab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031187.4 | NP_112464.4 | 2 - 3 | 428 | 61 | |
| GenBank mRNA | BC011328.1 | - | 2 - 3 | 393 | 3994444 | 61 |
| L00653.1 | - | 2 - 3 | 259 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tpsab1
Genomic Map