Assay Details
| Gene Symbol: Tpsab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031187.4 | NP_112464.4 | 3 - 4 | 691 | 140 | |
| NR_033521.1 | 3 - 4 | 593 | 140 | |||
| GenBank mRNA | BC011328.1 | - | 3 - 4 | 656 | 3994444 | 140 |
| L00653.1 | - | 3 - 4 | 522 | 140 | ||
| U42405.1 | - | 3 - 4 | 410 | 140 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tpsab1
Genomic Map