| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300010122.mm.1 |
NM_007378.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010122.mm.1 |
NM_007378.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010122.mm.1 |
TC0300001242.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300010124.mm.1 |
NM_007378.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010124.mm.1 |
NM_007378.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010124.mm.1 |
TC0300001242.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300010127.mm.1 |
NM_007378.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010127.mm.1 |
NM_007378.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010127.mm.1 |
TC0300001242.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300010125.mm.1 |
NM_007378.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010125.mm.1 |
NM_007378.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010125.mm.1 |
TC0300001242.mm.1 |
MTA TC |