Assay Details
| Gene Symbol: Dach1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038610.2 | NP_001033699.1 | 2 - 3 | 1396 | 152 | |
| NM_007826.3 | NP_031852.1 | 2 - 3 | 1396 | 152 | ||
| GenBank mRNA | AF090436.1 | - | 2 - 3 | 940 | 152 | |
| AF090437.1 | - | 2 - 3 | 940 | 152 | ||
| AF102547.1 | - | 2 - 3 | 940 | 152 | ||
| AF129510.1 | - | 2 - 3 | 1158 | 152 | ||
| AJ005669.1 | - | 2 - 3 | 458 | 152 | ||
| AK047409.1 | - | 2 - 3 | 262 | 152 | ||
| AK053594.1 | - | 2 - 3 | 605 | 152 | ||
| AK139060.1 | - | 2 - 3 | 1397 | 152 | ||
| BC059934.1 | - | 2 - 3 | 1174 | 152 | ||
| BC078644.1 | - | 2 - 3 | 930 | 152 | ||
| BC141130.1 | - | 2 - 3 | 1002 | 9055742 | 152 | |
| BC145440.1 | - | 2 - 3 | 1002 | 152 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dach1
Genomic Map