Assay Details
| Gene Symbol: Diap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK140734.1 | - | 5 - 6 | 592 | 65 | |
| AK220466.1 | - | 5 - 6 | 601 | 65 | ||
| BC021396.1 | - | 4 - 5 | 631 | 65 | ||
| BC070412.1 | - | 6 - 7 | 766 | 5705112 | 65 | |
| U96963.1 | - | 4 - 5 | 604 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Diap1
Genomic Map