Assay Details
| Gene Symbol: Diap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK140734.1 | - | 9 - 10 | 986 | 86 | |
| AK220466.1 | - | 9 - 10 | 995 | 86 | ||
| BC070412.1 | - | 10 - 11 | 1160 | 5705112 | 86 | |
| U96963.1 | - | 8 - 9 | 998 | 86 | ||
More Information
Set Membership: |
|

Diap1
Genomic Map