Assay Details
| Gene Symbol: Diap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK140734.1 | - | 10 - 11 | 1096 | 103 | |
| AK220466.1 | - | 10 - 11 | 1105 | 103 | ||
| BC021396.1 | - | 9 - 10 | 1135 | 103 | ||
| BC070412.1 | - | 11 - 12 | 1270 | 5705112 | 103 | |
| U96963.1 | - | 9 - 10 | 1108 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Diap1
Genomic Map