Assay Details
| Gene Symbol: Diap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK135915.1 | - | 2 - 3 | 275 | 56 | |
| AK156391.1 | - | 13 - 14 | 1859 | 56 | ||
| AK220466.1 | - | 26 - 27 | 3608 | 56 | ||
| BC070412.1 | - | 27 - 28 | 3773 | 5705112 | 56 | |
| U96963.1 | - | 25 - 26 | 3611 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Diap1
Genomic Map