Assay Details
| Gene Symbol: Dlg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109752.1 | NP_001103222.1 | 17 - 18 | 2165 | 72 | |
| NM_007864.3 | NP_031890.1 | 17 - 18 | 2174 | 72 | ||
| GenBank mRNA | BC014807.1 | - | 17 - 18 | 1910 | 4501403 | 72 |
| D50621.1 | - | 17 - 18 | 1916 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dlg4
Genomic Map