Assay Details
| Gene Symbol: Dlg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109752.1 | NP_001103222.1 | 18 - 19 | 2277 | 77 | |
| NM_007864.3 | NP_031890.1 | 18 - 19 | 2286 | 77 | ||
| GenBank mRNA | AK028242.1 | - | 1 - 2 | 112 | 77 | |
| BC014807.1 | - | 18 - 19 | 2022 | 4501403 | 77 | |
| D50621.1 | - | 18 - 19 | 2028 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dlg4
Genomic Map