Assay Details
| Gene Symbol: Phc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042623.2 | NP_001036088.1 | 3 - 4 | 325 | 67 | |
| NM_001271579.1 | NP_001258508.1 | 3 - 4 | 296 | 67 | ||
| NM_007905.3 | NP_031931.2 | 3 - 4 | 325 | 67 | ||
| GenBank mRNA | AK036370.1 | - | 3 - 4 | 285 | 67 | |
| AK132756.1 | - | 3 - 4 | 283 | 67 | ||
| AK167590.1 | - | 3 - 4 | 314 | 67 | ||
| BC046535.1 | - | 3 - 4 | 306 | 4935713 | 67 | |
| BC052394.1 | - | 3 - 4 | 658 | 5719837 | 67 | |
| BU610858.1 | - | 3 - 4 | 296 | 67 | ||
| CJ130893.1 | - | 3 - 4 | 325 | 67 | ||
| S73882.1 | - | 3 - 4 | 281 | 67 | ||
| U63386.1 | - | 2 - 3 | 247 | 67 | ||
More Information
Set Membership: |
|

Phc1
Genomic Map