Assay Details
| Gene Symbol: Phc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042623.2 | NP_001036088.1 | 8 - 9 | 2007 | 67 | |
| NM_001271579.1 | NP_001258508.1 | 9 - 10 | 2134 | 67 | ||
| NM_007905.3 | NP_031931.2 | 9 - 10 | 2163 | 67 | ||
| NR_073364.1 | 9 - 10 | 2170 | 67 | |||
| GenBank mRNA | AK036370.1 | - | 9 - 10 | 2123 | 67 | |
| AK132756.1 | - | 8 - 9 | 1965 | 67 | ||
| AK167590.1 | - | 8 - 9 | 1996 | 67 | ||
| BC046535.1 | - | 9 - 10 | 2150 | 4935713 | 67 | |
| BC052394.1 | - | 9 - 10 | 2496 | 5719837 | 67 | |
| S73882.1 | - | 9 - 10 | 2125 | 67 | ||
| S73883.1 | - | 9 - 10 | 2132 | 67 | ||
| U63386.1 | - | 8 - 9 | 2091 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Phc1
Genomic Map